missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEMA6B Partial ORF (NP_064626, 28 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010501-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16162666

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the semaphorin family, a group of proteins characterized by the presence of a conserved semaphorin (sema) domain. Whereas some semaphorins are transmembrane proteins, others are secreted. Semaphorins play a major role in axon guidance. The protein encoded by this gene may be involved in both peripheral and central nervous system development. [provided by RefSeq]

Sequence: PEEPPPLSVAPRDYLNHYPVFVGSGPGRLTPAEGADDLNIQRVLRVNRTLFIGDRDNLYRVELEPPTSTELRYQRKLTWRSNPSDINVCRMKGKQEGEC
Specifications

Specifications

NP_064626
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PEEPPPLSVAPRDYLNHYPVFVGSGPGRLTPAEGADDLNIQRVLRVNRTLFIGDRDNLYRVELEPPTSTELRYQRKLTWRSNPSDINVCRMKGKQEGEC
RUO
SEMA6B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10501
SEMA6B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SEM-SEMA-Y/SEMA-VIB/SEMAN/semaZ
SEMA6B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers