Learn More
Abnova™ Human SEMA7A Partial ORF (NP_003603, 536 a.a. - 633 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008482-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene binds to cell surfaces through a glycosylphosphatidylinositol (GPI) linkage. The encoded glycoprotein is found on activated lymphocytes and erythrocytes. This protein may be involved in immunomodulatory and neuronal processes. Defects in this gene can result in loss of bone mineral density (BMD). Three transcript variants encoding different isoforms have been found for this gene
Sequence: EPHKECPNPKPDKAPLQKVSLAPNSRYYLSCPMESRHATYSWRHKENVEQSCEPGHQSPNCILFIENLTAQQYGHYFCEAQEGSYFREAQHWQLLPEDSpecifications
NP_003603 | |
Liquid | |
8482 | |
SEMA7A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD108/CDw108/H-SEMA-K1/H-Sema-L/JMH/MGC126692/MGC126696/SEMAK1/SEMAL | |
SEMA7A | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPHKECPNPKPDKAPLQKVSLAPNSRYYLSCPMESRHATYSWRHKENVEQSCEPGHQSPNCILFIENLTAQQYGHYFCEAQEGSYFREAQHWQLLPED | |
RUO | |
SEMA7A | |
Wheat Germ (in vitro) | |
GST |