Learn More
Abnova™ Human SENP8 Full-length ORF (NP_660205.2, 1 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00123228-P01.10ug
Additional Details : Weight : 0.00010kg
Description
NEDD8 (MIM 603171) is a ubiquitin-like protein that becomes conjugated to the cullin (see CUL1; MIM 603134) subunit of several ubiquitin ligases. This conjugation, called neddylation, is required for optimal ubiquitin ligase activity. NEDD8-specific deneddylases, such as NEDP1, or DEN1, are required to process the NEDD8 propeptide at a C-terminal diglycine motif and to remove NEDD8 from cullins (Gan-Erdene et al., 2003 [PubMed 12759363]).[supplied by OMIM]
Sequence: MDPVVLSYMDSLLRQSDVSLLDPPSWLNDHIIGFAFEYFANSQFHDCSDHVSFISPEVTQFIKCTSNPAEIAMFLEPLDLPNKRVVFLAINDNSNQAAGGTHWSLLVYLQDKNSFFHYDSHSRSNSVHAKQVAEKLEAFLGRKGDKLAFVEEKAPAQQNSYDCGMYVICNTEALCQNFFRQQTESLLQLLTPAYITKKRGEWKDLIATLAKKSpecifications
NP_660205.2 | |
Liquid | |
123228 | |
SENP8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDPVVLSYMDSLLRQSDVSLLDPPSWLNDHIIGFAFEYFANSQFHDCSDHVSFISPEVTQFIKCTSNPAEIAMFLEPLDLPNKRVVFLAINDNSNQAAGGTHWSLLVYLQDKNSFFHYDSHSRSNSVHAKQVAEKLEAFLGRKGDKLAFVEEKAPAQQNSYDCGMYVICNTEALCQNFFRQQTESLLQLLTPAYITKKRGEWKDLIATLAKK | |
RUO | |
SENP8 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEN1/HsT17512/NEDP1/PRSC2 | |
SENP8 | |
Yes | |
wheat germ expression system |