Learn More
Abnova™ Human SEPT11 Full-length ORF (NP_060713.1, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055752-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking (Hanai et al., 2004 [PubMed 15196925]; Nagata et al., 2004 [PubMed 15485874]).[supplied by OMIM]
Sequence: MAVAVGRPSNEELRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYIDAQFEAYLQEELKIKRSLFNYHDTRIHACLYFIAPTGHSLKSLDLVTMKKLDSKVNIIPIIAKADTIAKNELHKFKSKIMSELVSNGVQIYQFPTDEETVAEINATMSVHLPFAVVGSTEEVKIGNKMAKARQYPWGVVQVENENHCDFVKLREMLIRVNMEDLREQTHTRHYELYRRCKLEEMGFKDTDPDSKPFSLQETYEAKRNEFLGELQKKEEEMRQMFVMRVKEKEAELKEAEKELHEKFDLLKRTHQEEKKKVEDKKKELEEEVNNFQKKKAAAQLLQSQAQQSGAQQTKKDKDKKNASFTSpecifications
NP_060713.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEPT11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55752 | |
SEPT11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVAVGRPSNEELRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYIDAQFEAYLQEELKIKRSLFNYHDTRIHACLYFIAPTGHSLKSLDLVTMKKLDSKVNIIPIIAKADTIAKNELHKFKSKIMSELVSNGVQIYQFPTDEETVAEINATMSVHLPFAVVGSTEEVKIGNKMAKARQYPWGVVQVENENHCDFVKLREMLIRVNMEDLREQTHTRHYELYRRCKLEEMGFKDTDPDSKPFSLQETYEAKRNEFLGELQKKEEEMRQMFVMRVKEKEAELKEAEKELHEKFDLLKRTHQEEKKKVEDKKKELEEEVNNFQKKKAAAQLLQSQAQQSGAQQTKKDKDKKNASFT | |
RUO | |
SEPT11 | |
Recombinant | |
wheat germ expression system |