missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SEPT11 Full-length ORF (NP_060713.1, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055752-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121763

  • 4515.00 SEK / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking (Hanai et al., 2004 [PubMed 15196925]; Nagata et al., 2004 [PubMed 15485874]).[supplied by OMIM]

Sequence: MAVAVGRPSNEELRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYIDAQFEAYLQEELKIKRSLFNYHDTRIHACLYFIAPTGHSLKSLDLVTMKKLDSKVNIIPIIAKADTIAKNELHKFKSKIMSELVSNGVQIYQFPTDEETVAEINATMSVHLPFAVVGSTEEVKIGNKMAKARQYPWGVVQVENENHCDFVKLREMLIRVNMEDLREQTHTRHYELYRRCKLEEMGFKDTDPDSKPFSLQETYEAKRNEFLGELQKKEEEMRQMFVMRVKEKEAELKEAEKELHEKFDLLKRTHQEEKKKVEDKKKELEEEVNNFQKKKAAAQLLQSQAQQSGAQQTKKDKDKKNASFT
Specifications

Specifications

NP_060713.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SEPT11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
55752
SEPT11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVAVGRPSNEELRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYIDAQFEAYLQEELKIKRSLFNYHDTRIHACLYFIAPTGHSLKSLDLVTMKKLDSKVNIIPIIAKADTIAKNELHKFKSKIMSELVSNGVQIYQFPTDEETVAEINATMSVHLPFAVVGSTEEVKIGNKMAKARQYPWGVVQVENENHCDFVKLREMLIRVNMEDLREQTHTRHYELYRRCKLEEMGFKDTDPDSKPFSLQETYEAKRNEFLGELQKKEEEMRQMFVMRVKEKEAELKEAEKELHEKFDLLKRTHQEEKKKVEDKKKELEEEVNNFQKKKAAAQLLQSQAQQSGAQQTKKDKDKKNASFT
RUO
SEPT11
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers