missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEPT12 Full-length ORF (AAH35619.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH35619.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 124404 |
Molecular Weight (g/mol) | 67.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137593
|
Abnova™
H00124404-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16127593
|
Abnova™
H00124404-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).[supplied by OMIM]
Sequence: MDPLRRSPSPCLSSQPSSPSTPPCEMLGPVGIEAVLDQLKIKAMKMGFEFNIMVVGQSGLGKSTMVNTLFKSKVWKSNPPGLGVPTPQTLQLHSLTHVIEEKGVKLKLTVTDTPGFGDQINNDNCWDPILGYINEQYEQYLQEEILITRQRHIPDTRVHCCVYFVPPTGHCLRPLDIEFLQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFDEDINDKILNSKLRDRIPFAVVGADQEHLVNGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEFSpecifications
AAH35619.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDPLRRSPSPCLSSQPSSPSTPPCEMLGPVGIEAVLDQLKIKAMKMGFEFNIMVVGQSGLGKSTMVNTLFKSKVWKSNPPGLGVPTPQTLQLHSLTHVIEEKGVKLKLTVTDTPGFGDQINNDNCWDPILGYINEQYEQYLQEEILITRQRHIPDTRVHCCVYFVPPTGHCLRPLDIEFLQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFDEDINDKILNSKLRDRIPFAVVGADQEHLVNGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEF | |
RUO | |
SEPT12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
124404 | |
SEPT12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25410 | |
SEPT12 | |
Recombinant | |
wheat germ expression system |