Learn More
Abnova™ Human SFMBT1 Full-length ORF (NP_001005158.1, 1 a.a. - 866 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051460-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition. Several alternative splice variants have been characterized. [provided by RefSeq]
Sequence: MNGEQQLDADAGSGMEEVELSWEDYLEETGSTAVPYGSFKHVDTRLQNGFAPGMKLEVAVRTDPETYWVATVITTCEQLLLLRYDGYGEDRRADFWCDIRKADLYPIGWCEQNKKTLEAPEGIRDKVSDWDEFLRQTLIGACSPPVPLLEGLRNGRNPLDLIAPGSRLECQAFQDSLSTWIVTVVENIGGRLKLRYEGLESSDNYEHWLYYLDPFLHHVGWAAQQGYELQPPSAIRHLKNEAEWQEILAKVKEEEEEPLPSYLFKDKQVIGIHTFSVNMKLEAVDPWSPFGISPATVVKVFDEKYFLVEMDDLRPENHARRSFVCHADSPGIFPVQWSLKNGLHISPPPGYPSQDFDWADYLKQCGAEAAPQRCFPPLISEHEFKENMKLEAVNPILPEEVCVATITAVRGSYLWLQLEGSKKPIPECIVSVESMDIFPLGWCETNGHPLSTPRRARVYKQRKIAVVQPEKQVPSSRTVHEGLRNQELNSTESVMINGKYCCPKIYFNHRCFSGPYLNKGRIAELPQCVGPGNCVLVLREVLTLLINAAYKPSRVLRELQLDKDSVWHGCGEVLKAKYKGKSYRATVEIVKTADRVTEFCRQTCIKLECCPNLFGPRMVLDKCSENCSVLTKTKYTHYYGKKKNKRIGRPPGGHSNLACALKKASKRRKRRKNVFVHKKKRSSASVDNTPAGSPQGSGGEDEDDPDEGDDDSLSEGSTSEQQDELQEESEMSEKKSCSSSPTQSEISTSLPPDRQRRKRELRTFSFSDDENKPPSPKEIRIEVAERLHLDSNPLKWSVADVVRFIRSTDCAPLARIFLDQEIDGQALLLLTLPTVQECMDLKLGPAIKLCHHIERIKFAFYEQFANSpecifications
NP_001005158.1 | |
Liquid | |
51460 | |
SFMBT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNGEQQLDADAGSGMEEVELSWEDYLEETGSTAVPYGSFKHVDTRLQNGFAPGMKLEVAVRTDPETYWVATVITTCEQLLLLRYDGYGEDRRADFWCDIRKADLYPIGWCEQNKKTLEAPEGIRDKVSDWDEFLRQTLIGACSPPVPLLEGLRNGRNPLDLIAPGSRLECQAFQDSLSTWIVTVVENIGGRLKLRYEGLESSDNYEHWLYYLDPFLHHVGWAAQQGYELQPPSAIRHLKNEAEWQEILAKVKEEEEEPLPSYLFKDKQVIGIHTFSVNMKLEAVDPWSPFGISPATVVKVFDEKYFLVEMDDLRPENHARRSFVCHADSPGIFPVQWSLKNGLHISPPPGYPSQDFDWADYLKQCGAEAAPQRCFPPLISEHEFKENMKLEAVNPILPEEVCVATITAVRGSYLWLQLEGSKKPIPECIVSVESMDIFPLGWCETNGHPLSTPRRARVYKQRKIAVVQPEKQVPSSRTVHEGLRNQELNSTESVMINGKYCCPKIYFNHRCFSGPYLNKGRIAELPQCVGPGNCVLVLREVLTLLINAAYKPSRVLRELQLDKDSVWHGCGEVLKAKYKGKSYRATVEIVKTADRVTEFCRQTCIKLECCPNLFGPRMVLDKCSENCSVLTKTKYTHYYGKKKNKRIGRPPGGHSNLACALKKASKRRKRRKNVFVHKKKRSSASVDNTPAGSPQGSGGEDEDDPDEGDDDSLSEGSTSEQQDELQEESEMSEKKSCSSSPTQSEISTSLPPDRQRRKRELRTFSFSDDENKPPSPKEIRIEVAERLHLDSNPLKWSVADVVRFIRSTDCAPLARIFLDQEIDGQALLLLTLPTVQECMDLKLGPAIKLCHHIERIKFAFYEQFAN | |
RUO | |
SFMBT1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
124.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434L243/RU1/SFMBT | |
SFMBT1 | |
Yes | |
wheat germ expression system |