Learn More
Abnova™ Human SGCD Partial ORF (NP_000328.2, 114 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_000328.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6444 |
Molecular Weight (g/mol) | 34.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115175
|
Abnova™
H00006444-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16105175
|
Abnova™
H00006444-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is one of the four known components of the sarcoglycan complex, which is a subcomplex of the dystrophin-glycoprotein complex (DGC). DGC forms a link between the F-actin cytoskeleton and the extracellular matrix. This protein is expressed most abundantly in skeletal and cardiac muscle. Mutations in this gene have been associated with autosomal recessive limb-girdle muscular dystrophy and dilated cardiomyopathy. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq]
Sequence: ILNDQTKVLTQLITGPKAVEAYGKKFEVKTVSGKLLFSADNNEVVVGAERLRVLGAEGTVFPKSIETPNVRADPFKSpecifications
NP_000328.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
35DAG/CMD1L/DAGD/MGC22567/SG-delta/SGCDP/SGD | |
SGCD | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6444 | |
SGCD (Human) Recombinant Protein (Q01) | |
ILNDQTKVLTQLITGPKAVEAYGKKFEVKTVSGKLLFSADNNEVVVGAERLRVLGAEGTVFPKSIETPNVRADPFK | |
RUO | |
SGCD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |