Learn More
Abnova™ Human SGIP1 Full-length ORF (AAH40516.1, 1 a.a. - 859 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084251-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SGIP1 functions as an endocytic protein that affects signaling by receptors in neuronal systems involved in energy homeostasis via its interaction with endophilins (see SH3GL3; MIM 603362) (Trevaskis et al., 2005 [PubMed 15919751] and Uezu et al., 2007 [PubMed 17626015]).[supplied by OMIM]
Sequence: MMEGLKKRTRKAFGIRKKEKDTDSTGSPDRDGIQGKKKTQKTQLLLTSFFWLRALSLTLSQKKSNGAPNGFYAEIDWERYNSPELDEEGYSIRPEEPGSTKGKHFYSSSESEEEEESHKKFNIKIKPLQSKDILKNAATVDELKASIGNIALSPSPVRKSPRRSQGAIKRNLSSEEVARPRRSTPTPELISKKPPDDTTALAPLFGPPLESAFDEQKTEVLLDQPEIWGSGQPINPSMESPKLTRPFPTGTPPPLPPKNVPATPPRTGSPLTIGPGNDQSATEVKIEKLPSINDLDSIFGPVLSPKSVAVNAEEKWVHFSDTSPEHVTPELTPREKVVSPPATPDNPADSPAPGPLGPPGPTGPPGPPGPPRNVLSPLNLEEVQKKVAEQTFIKDDYLETISSPKDFGLGQRATPPPPPPPTYRTVVSSPGPGSGPGPGTTSDGKTEAQRYQVICPSLQAGGNELDSYSGASSPARPATPLVPCRSTTPPPPPPRPPSRPKLPPGKPGVGDVSRPFSPPIHSSSPPPIAPLARAESTSSISSTNSLSAATTPTVENEQPSLVWFDRGKFYLTFEGSSRGPSPLTMGAQDTLPVAAAFTETVNAYFEGADPSKCIVKITGEMVLSFPAGITRHFANNPSPAALTFRVINFSRLEHVLPNPQLLCCDNTQNDANTKEFWVNMPNLMTHLKKVSEQKPQATYYNVDMLKYQVSAQGIQSTPLNLAVNWRCEPSSTDLRIDYKYNTDAMTTAVALNNVQFLVPIDGGVTKLQAVLPPAVWNAEQQRILWKIPDISQKSENGGVGSLLARFQLSEGPSKPSPLVVQFTSEGSTLSGCDIELVGAGYRFSLIKKRFAAGKYLADNSpecifications
AAH40516.1 | |
Liquid | |
84251 | |
SGIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMEGLKKRTRKAFGIRKKEKDTDSTGSPDRDGIQGKKKTQKTQLLLTSFFWLRALSLTLSQKKSNGAPNGFYAEIDWERYNSPELDEEGYSIRPEEPGSTKGKHFYSSSESEEEEESHKKFNIKIKPLQSKDILKNAATVDELKASIGNIALSPSPVRKSPRRSQGAIKRNLSSEEVARPRRSTPTPELISKKPPDDTTALAPLFGPPLESAFDEQKTEVLLDQPEIWGSGQPINPSMESPKLTRPFPTGTPPPLPPKNVPATPPRTGSPLTIGPGNDQSATEVKIEKLPSINDLDSIFGPVLSPKSVAVNAEEKWVHFSDTSPEHVTPELTPREKVVSPPATPDNPADSPAPGPLGPPGPTGPPGPPGPPRNVLSPLNLEEVQKKVAEQTFIKDDYLETISSPKDFGLGQRATPPPPPPPTYRTVVSSPGPGSGPGPGTTSDGKTEAQRYQVICPSLQAGGNELDSYSGASSPARPATPLVPCRSTTPPPPPPRPPSRPKLPPGKPGVGDVSRPFSPPIHSSSPPPIAPLARAESTSSISSTNSLSAATTPTVENEQPSLVWFDRGKFYLTFEGSSRGPSPLTMGAQDTLPVAAAFTETVNAYFEGADPSKCIVKITGEMVLSFPAGITRHFANNPSPAALTFRVINFSRLEHVLPNPQLLCCDNTQNDANTKEFWVNMPNLMTHLKKVSEQKPQATYYNVDMLKYQVSAQGIQSTPLNLAVNWRCEPSSTDLRIDYKYNTDAMTTAVALNNVQFLVPIDGGVTKLQAVLPPAVWNAEQQRILWKIPDISQKSENGGVGSLLARFQLSEGPSKPSPLVVQFTSEGSTLSGCDIELVGAGYRFSLIKKRFAAGKYLADN | |
RUO | |
SGIP1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A16142/DKFZp761D221/FLJ33378/FLJ43054 | |
SGIP1 | |
Yes | |
wheat germ expression system |