missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SGK2 Full-length ORF (NP_733794.1, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_733794.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10110 |
Molecular Weight (g/mol) | 67.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124512
|
Abnova™
H00010110-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16114512
|
Abnova™
H00010110-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a serine/threonine protein kinase. Although this gene product is similar to serum- and glucocorticoid-induced protein kinase (SGK), this gene is not induced by serum or glucocorticoids. This gene is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Two alternate transcripts encoding two different isoforms have been described. [provided by RefSeq]
Sequence: MNSSPAGTPSPQPSRANGNINLGPSANPNAQPTDFDFLKVIGKGNYGKVLLAKRKSDGAFYAVKVLQKKSILKKKEQSHIMAERSVLLKNVRHPFLVGLRYSFQTPEKLYFVLDYVNGGELFFHLQRERRFLEPRARFYAAEVASAIGYLHSLNIIYRDLKPENILLDCQGHVVLTDFGLCKEGVEPEDTTSTFCGTPEYLAPEVLRKEPYDRAVDWWCLGAVLYEMLHGLPPFYSQDVSQMYENILHQPLQIPGGRTVAACDLLQSLLHKDQRQRLGSKADFLEIKNHVFFSPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPDTVASSSGASSAFLGFSYAPEDDDILDCSpécification
NP_733794.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNSSPAGTPSPQPSRANGNINLGPSANPNAQPTDFDFLKVIGKGNYGKVLLAKRKSDGAFYAVKVLQKKSILKKKEQSHIMAERSVLLKNVRHPFLVGLRYSFQTPEKLYFVLDYVNGGELFFHLQRERRFLEPRARFYAAEVASAIGYLHSLNIIYRDLKPENILLDCQGHVVLTDFGLCKEGVEPEDTTSTFCGTPEYLAPEVLRKEPYDRAVDWWCLGAVLYEMLHGLPPFYSQDVSQMYENILHQPLQIPGGRTVAACDLLQSLLHKDQRQRLGSKADFLEIKNHVFFSPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPDTVASSSGASSAFLGFSYAPEDDDILDC | |
RUO | |
SGK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10110 | |
SGK2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
H-SGK2/dJ138B7.2 | |
SGK2 | |
Recombinant | |
wheat germ expression system |