missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human Sharpin Partial ORF (NP_112236, 51 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081858-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16130447

  • 5030.41 SEK / 25µg

Estimated Shipment: 14-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: ERPGRFRLELLGAGPGAVNLEWPLESVSYTIRGPTQHELQPPPGGPGTLSLHFLNPQEAQRWAVLVRGATVEGQNGSKSNSPPALGPEACPVSLPSPP
Specifications

Specifications

NP_112236
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ERPGRFRLELLGAGPGAVNLEWPLESVSYTIRGPTQHELQPPPGGPGTLSLHFLNPQEAQRWAVLVRGATVEGQNGSKSNSPPALGPEACPVSLPSPP
RUO
SHARPIN
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
81858
Sharpin (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434N1923/SIPL1
SHARPIN
Yes
Videos
SDS
Documents
Certificates