missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SHC2 Partial ORF (XP_375550, 718 a.a. - 829 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025759-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195056

  • 6845.00 SEK / 25µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LALTQPCALTALDQGPSPSLRDACSLPWDVGSTGTAPPGDGYVQADARGPPDHEEHLYVNTQGLDAPEPEDSPKKDLFDMRPFEDALKLHECSVAAGVTAAPLPLEDQWPSP
Specifications

Specifications

XP_375550
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.06kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LALTQPCALTALDQGPSPSLRDACSLPWDVGSTGTAPPGDGYVQADARGPPDHEEHLYVNTQGLDAPEPEDSPKKDLFDMRPFEDALKLHECSVAAGVTAAPLPLEDQWPSP
RUO
SHC2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
25759
SHC2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCK/SHCB/SLI
SHC2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers