Learn More
Abnova™ Human SIAHBP1 Full-length ORF (AAH11979, 1 a.a. - 516 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH11979 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22827 |
Molecular Weight (g/mol) | 82.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16126632
|
Abnova™
H00022827-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16116632
|
Abnova™
H00022827-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene. [provided by RefSeq]
Sequence: MENGQSTAAKLGLPPLTPEQQEALQKAKKYAMEQSIKSVLVKQTIAHQQQQLTNLQMAAVTMGFGDPLSPLQSMAAQRQRALAIMCRVYVGSIYYELGEDTIRQAFAPFGPIKSIDMSWDSVTMKHKGFAFVEYEVPEAAQLALEQMNSVMLGGRNIKVGRPSNIGQAQPIIDQLAEEARAFNRIYVASVHQDLSDDDIKSVFEAFGKIKSCTLARDPTTGKHKGYGFIEYEKAQSSQDAVSSMNLFDLGGQYLRVGKAVTPPMPLLTPATPGGLPPAAAVAAAAATAKITAQEAVAGAAVLGTLGTPGLVSPALTLAQPLGTLPQAVMAAQAPGVITGVTPARPPIPVTIPSVGVVNPILASPPTLGLLEPKKEKEEEELFPESERPEMLSEQEHMSISGSSARHMVMQKLLRKQESTVMVLRNMVDPKDIDDDLEGEVTEECGKFGAVNRVIIYQEKQGEEEDAEIIVKIFVEFSIASETHKAIQALNGRWFAGRKVVAEVYDQERFDNSDLSASpecifications
AAH11979 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MENGQSTAAKLGLPPLTPEQQEALQKAKKYAMEQSIKSVLVKQTIAHQQQQLTNLQMAAVTMGFGDPLSPLQSMAAQRQRALAIMCRVYVGSIYYELGEDTIRQAFAPFGPIKSIDMSWDSVTMKHKGFAFVEYEVPEAAQLALEQMNSVMLGGRNIKVGRPSNIGQAQPIIDQLAEEARAFNRIYVASVHQDLSDDDIKSVFEAFGKIKSCTLARDPTTGKHKGYGFIEYEKAQSSQDAVSSMNLFDLGGQYLRVGKAVTPPMPLLTPATPGGLPPAAAVAAAAATAKITAQEAVAGAAVLGTLGTPGLVSPALTLAQPLGTLPQAVMAAQAPGVITGVTPARPPIPVTIPSVGVVNPILASPPTLGLLEPKKEKEEEELFPESERPEMLSEQEHMSISGSSARHMVMQKLLRKQESTVMVLRNMVDPKDIDDDLEGEVTEECGKFGAVNRVIIYQEKQGEEEDAEIIVKIFVEFSIASETHKAIQALNGRWFAGRKVVAEVYDQERFDNSDLSA | |
RUO | |
PUF60 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
22827 | |
SIAHBP1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FIR/FLJ31379/RoBPI/SIAHBP1 | |
PUF60 | |
Recombinant | |
wheat germ expression system |