missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SIGLEC10 Partial ORF (NP_149121, 589 a.a. - 696 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_149121 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 89790 |
Molecular Weight (g/mol) | 37.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121057
|
Abnova™
H00089790-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131057
|
Abnova™
H00089790-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6 (MIM 604405).[supplied by OMIM]
Sequence: SRHSTILDYINVVPTAGPLAQKRNQKATPNSPRTPLPPGAPSPESKKNQKKQYQLPSFPEPKSSTQAPESQESQEELHYATLNFPGVRPRPEARMPKGTQADYAEVKFSpecifications
NP_149121 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126774/PRO940/SIGLEC-10/SLG2 | |
SIGLEC10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
89790 | |
SIGLEC10 (Human) Recombinant Protein (Q01) | |
SRHSTILDYINVVPTAGPLAQKRNQKATPNSPRTPLPPGAPSPESKKNQKKQYQLPSFPEPKSSTQAPESQESQEELHYATLNFPGVRPRPEARMPKGTQADYAEVKF | |
RUO | |
SIGLEC10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |