missing translation for 'onlineSavingsMsg'
Läs mer

Gibco™ Human IL-15 Receptor alpha (soluble) Fc Recombinant Protein, PeproTech®

Produktkod. 17829575
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
100 μg
20 μg
250 μg
500 μg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 17829575

Brand: Gibco™ 20015RA1MG

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Recombinant Human sIL-15 Receptor alpha Fc has a calculated molecular weight of 44.4 kDa protein and contains 408 amino acid residues, which consists of the extracellular domain of IL-15R alpha fused to the Fc portion of human IgG. Due to glycosylation, it migrates at an apparent molecular weight of 50-60 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer Q13261
För användning med (applikation) Functional Assay
Formatera Lyophilized
Formulering protein with no preservative
Gen-ID (Entrez) 3601
Molekylvikt (g/mol) 44.4 kDa
Namn Human IL-15 Receptor alpha (soluble) Fc
Reningsmetod HPLC, SDS-PAGE
Kvantitet 1 mg
Regulatorisk status RUO
Endotoxinkoncentration <1 EU/μg
Gene Alias AA690181; CD215; IL 15 receptor; IL 15 receptor subunit a; IL 15 R; IL 15 R alpha; IL 15 R a; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit a; IL15R; IL15R alpha; IL15R a; Il15ra; IL-15 RA; IL-15 R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit a; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15 R; sIL-15 receptor subunit alpha; sIL15R; sIL-15 RA; sIL-15 R-alpha; soluble IL 15 R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha
Vanligt namn IL15RA
Gensymbol IL15RA
Biologisk aktivitet Determined by its ability to inhibit the proliferation of mouse CTLL-2 cells in the presence of Recombinant Human IL-15.
Produkttyp Protein
Konjugera Unconjugated
Rekombinant Recombinant
Protein Tag Fc-tag
Sekvens ITCPPPMSVE HADIWVKSYS LYSRERYICN SGFKRKAGTS SLTECVLNKA TNVAHWTTPS LKCIRDPALV HQRPAPPSTV TTAGVTPQPE SLSPSGKEPA ASSPSSNNTA ATTAAIVPGS QLMPSKSPST GTTEISSHES SHGTPSQTTA KNWELTASAS HQPPGVYPQG HSDTTGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGK
Innehåll och lagring -20°C
Aktivitet Determined by its ability to inhibit the proliferation of mouse CTLL-2 cells in the presence of Recombinant Human IL-15.
Uttryckssystem Insect cells
Form Lyophilized
Renhet eller kvalitetsklass ≥98% by SDS-PAGE gel and HPLC analyses.
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.