missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Sin1/MAPKAP1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8168
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Sin1/MAPKAP1 Monoclonal specifically detects Sin1/MAPKAP1 in Human samples. It is validated for Western Blot.
Specifikationer
| Sin1/MAPKAP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| Q9BPZ7 | |
| MAPKAP1 | |
| E. coli-derived recombinant human Sin1/MAPKAP1 Lys408-Gln522 Accession # Q9BPZ7 | |
| 100 μg | |
| Primary | |
| Detects human Sin1/MAPKAP1 in ELISA. | |
| Human | |
| Purified |
| Western Blot | |
| 752219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| JC310, MAPKAP1, MEKK2-interacting protein 1, MGC2745, MIP1, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79109 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering