Learn More
Abnova™ Human SIRT4 Partial ORF (NP_036372.1, 215 a.a. - 314 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023409-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family. [provided by RefSeq]
Sequence: FQVPTCVQCGGHLKPDVVFFGDTVNPDKVDFVHKRVKEADSLLVVGSSLQVYSGYRFILTAWEKKLPIAILNIGPTRSDDLACLKLNSRCGELLPLIDPCSpecifications
NP_036372.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FQVPTCVQCGGHLKPDVVFFGDTVNPDKVDFVHKRVKEADSLLVVGSSLQVYSGYRFILTAWEKKLPIAILNIGPTRSDDLACLKLNSRCGELLPLIDPC | |
RUO | |
SIRT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23409 | |
SIRT4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC130046/MGC130047/MGC57437/SIR2L4 | |
SIRT4 | |
Recombinant | |
wheat germ expression system |