Läs mer
Gibco™ Human Skeletal Myoblasts (HSkM)
Human Skeletal Myoblasts
Beskrivning
Beskrivning
Primary Normal Human Skeletal Myoblasts (HSkM) are designed to undergo highly efficient differentiation directly following plating of cryopreserved cells. Each vial contains cells that have been cryopreserved in a medium containing 10% DMSO and will differentiate within 48 hours after plating. These cells do not require expansion prior to differentiation. Cell viability is >70% post thaw with a ≥50% myogenic index after 48 hours. Intended for use by researchers investigating the molecular and biochemical bases of various normal and disease processes. They are compatible with Bacmam delivery technologies for various downstream applications. Large size contains 4.5 x 10^6 ≤ N ≤ 7.5 x 10^6 cells per vial and small size contains 1.1 x 10^6 ≤ N ≤ 1.9 x 10^6 cells per vial.
Filmer
SDS
Dokument
Dokument
Produktspecifikationer