missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC12A6 Partial ORF (NP_005126, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009990-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121586

  • 6845.00 SEK / 25µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy. [provided by RefSeq]

Sequence: MPHFTVTKVEDPEEGAAASISQEPSLADIKARIQDSDEPDLSQNSITGEHSQLLDDGHKKARNAYLNNSNYEEGDEYFDKNLALFEEEMD
Specifications

Specifications

NP_005126
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MPHFTVTKVEDPEEGAAASISQEPSLADIKARIQDSDEPDLSQNSITGEHSQLLDDGHKKARNAYLNNSNYEEGDEYFDKNLALFEEEMD
RUO
SLC12A6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9990
SLC12A6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACCPN/DKFZp434D2135/KCC3/KCC3A/KCC3B
SLC12A6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers