missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC14A2 Partial ORF (NP_009094, 40 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_009094 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8170 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148275
|
Abnova™
H00008170-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138275
|
Abnova™
H00008170-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In mammalian cells, urea is the chief end-product of nitrogen catabolism and plays an important role in the urinary concentration mechanism. Thus, the plasma membrane of erythrocytes and some renal epithelial cells exhibit an elevated urea permeability that is mediated by highly selective urea transporters. In mammals, 2 urea transporters have been identified: the renal tubular urea transporter, UT2, and the erythrocyte urea transporter, UT11 (SLC14A1; MIM 111000).[supplied by OMIM]
Sequence: ALPLLEMPEEKDLRSSNEDSHIVKIEKLNERSKRKDDGVAHRDSAGQRCICLSKAVGYLTGDMKEYRIWLKDKHLALQFIDWVLRGTAQSpecifications
NP_009094 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16167/HUT2/MGC119566/MGC119567/UT-A2/UT2/UTA/UTR/hUT-A6 | |
SLC14A2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8170 | |
SLC14A2 (Human) Recombinant Protein (Q01) | |
ALPLLEMPEEKDLRSSNEDSHIVKIEKLNERSKRKDDGVAHRDSAGQRCICLSKAVGYLTGDMKEYRIWLKDKHLALQFIDWVLRGTAQ | |
RUO | |
SLC14A2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |