Läs mer
R&D Systems™ Human SLC16A4 (NP_004687.1) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | SLC16A4 |
| Tillträdesnummer | NP_004687.1 |
| Gen-ID (Entrez) | 9122 |
| Gene Alias | MCT 5, MCT 6, MCT5, MCT6, Monocarboxylate transporter 5, Monocarboxylic acid transporter 5, Monocarboxylic acid transporter 6, MOT5, SLC16A5, solute carrier family 16 (monocarboxylic acid transporters) member 5, Solute carrier family 16 member 5 |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Human |
| Kvantitet | 10 μg |