missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC18A3 Partial ORF (NP_003046, 476 a.a. - 532 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_003046 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6572 |
Molecular Weight (g/mol) | 32.01kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16125445
|
Abnova™
H00006572-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16115445
|
Abnova™
H00006572-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene. [provided by RefSeq]
Sequence: TRSRSERDVLLDEPPQGLYDAVRLRERPVSGQDGEPRSPPGPFDECEDDYNYYYTRSSpecifications
NP_003046 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.01kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12716/VACHT | |
SLC18A3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6572 | |
SLC18A3 (Human) Recombinant Protein (Q01) | |
TRSRSERDVLLDEPPQGLYDAVRLRERPVSGQDGEPRSPPGPFDECEDDYNYYYTRS | |
RUO | |
SLC18A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |