missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A38 Full-length ORF (AAH13194.1, 1 a.a. - 304 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054977-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16131003

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLC25A38 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]

Sequence: MIQNSRPSLLQPQDVGDTVETLMLHPVIKAFLCGSISGTCSTLLFQPLDLLKTRLQTLQPSDHGSRRVGMLAVLLKVVRTESLLGLWKGMSPSIVRCVPGVGIYFGTLYSLKQYFLRGHPPTALESVMLGVGSRSVAGVCMSPITVIKTRYESGKYGYESIYAALRSIYHSEGHRGLFSGLTATLLRDAPFSGIYLMFYNQTKNIVPHDQVDATLIPITNFSCGIFAGILASLVTQPADVIKTHMQLYPLKFQWIGQAVTLIFKDYGLRGFFQGGIPRALRRTLMAAMAWTVYEEMMAKMGLKS
Specifications

Specifications

AAH13194.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20551/FLJ22703
SLC25A38
Yes
wheat germ expression system
Liquid
54977
SLC25A38 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIQNSRPSLLQPQDVGDTVETLMLHPVIKAFLCGSISGTCSTLLFQPLDLLKTRLQTLQPSDHGSRRVGMLAVLLKVVRTESLLGLWKGMSPSIVRCVPGVGIYFGTLYSLKQYFLRGHPPTALESVMLGVGSRSVAGVCMSPITVIKTRYESGKYGYESIYAALRSIYHSEGHRGLFSGLTATLLRDAPFSGIYLMFYNQTKNIVPHDQVDATLIPITNFSCGIFAGILASLVTQPADVIKTHMQLYPLKFQWIGQAVTLIFKDYGLRGFFQGGIPRALRRTLMAAMAWTVYEEMMAKMGLKS
RUO
SLC25A38
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates