missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC25A46 Full-length ORF (NP_620128.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00091137-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SLC25A46 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]
Sequence: MHPRRPDGFDGLGYRGGARDEQGFGGAFPARSFSTGSDLGHWVTTPPDIPGSRNLHWGEKSPPYGVPTTSTPYEGPTEEPFSSGGGGSVQGQSSEQLNRFAGFGIGLASLFTENVLAHPCIVLRRQCQVNYHAQHYHLTPFTVINIMYSFNKTQGPRALWKGMGSTFIVQGVTLGAEGIISEFTPLPREVLHKWSPKQIGEHLLLKSLTYVVAMPFYSASLIETVQSEIIRDNTGILECVKEGIGRVIGMGVPHSKRLLPLLSLIFPTVLHGVLHYIISSVIQKFVLLILKRKTYNSHLAESTSPVQSMLDAYFPELIANFAASLCSDVILYPLETVLHRLHIQGTRTIIDNTDLGYEVLPINTQYEGMRDCINTIRQEEGVFGFYKGFGAVIIQYTLHAAVLQITKIIYSTLLQNNISpecifications
NP_620128.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC25A46 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
91137 | |
SLC25A46 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MHPRRPDGFDGLGYRGGARDEQGFGGAFPARSFSTGSDLGHWVTTPPDIPGSRNLHWGEKSPPYGVPTTSTPYEGPTEEPFSSGGGGSVQGQSSEQLNRFAGFGIGLASLFTENVLAHPCIVLRRQCQVNYHAQHYHLTPFTVINIMYSFNKTQGPRALWKGMGSTFIVQGVTLGAEGIISEFTPLPREVLHKWSPKQIGEHLLLKSLTYVVAMPFYSASLIETVQSEIIRDNTGILECVKEGIGRVIGMGVPHSKRLLPLLSLIFPTVLHGVLHYIISSVIQKFVLLILKRKTYNSHLAESTSPVQSMLDAYFPELIANFAASLCSDVILYPLETVLHRLHIQGTRTIIDNTDLGYEVLPINTQYEGMRDCINTIRQEEGVFGFYKGFGAVIIQYTLHAAVLQITKIIYSTLLQNNI | |
RUO | |
SLC25A46 | |
Recombinant | |
wheat germ expression system |