missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A46 Full-length ORF (NP_620128.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00091137-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136573

  • 3343.96 SEK / 10µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLC25A46 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]

Sequence: MHPRRPDGFDGLGYRGGARDEQGFGGAFPARSFSTGSDLGHWVTTPPDIPGSRNLHWGEKSPPYGVPTTSTPYEGPTEEPFSSGGGGSVQGQSSEQLNRFAGFGIGLASLFTENVLAHPCIVLRRQCQVNYHAQHYHLTPFTVINIMYSFNKTQGPRALWKGMGSTFIVQGVTLGAEGIISEFTPLPREVLHKWSPKQIGEHLLLKSLTYVVAMPFYSASLIETVQSEIIRDNTGILECVKEGIGRVIGMGVPHSKRLLPLLSLIFPTVLHGVLHYIISSVIQKFVLLILKRKTYNSHLAESTSPVQSMLDAYFPELIANFAASLCSDVILYPLETVLHRLHIQGTRTIIDNTDLGYEVLPINTQYEGMRDCINTIRQEEGVFGFYKGFGAVIIQYTLHAAVLQITKIIYSTLLQNNI
Specifications

Specifications

NP_620128.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLC25A46
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
91137
SLC25A46 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MHPRRPDGFDGLGYRGGARDEQGFGGAFPARSFSTGSDLGHWVTTPPDIPGSRNLHWGEKSPPYGVPTTSTPYEGPTEEPFSSGGGGSVQGQSSEQLNRFAGFGIGLASLFTENVLAHPCIVLRRQCQVNYHAQHYHLTPFTVINIMYSFNKTQGPRALWKGMGSTFIVQGVTLGAEGIISEFTPLPREVLHKWSPKQIGEHLLLKSLTYVVAMPFYSASLIETVQSEIIRDNTGILECVKEGIGRVIGMGVPHSKRLLPLLSLIFPTVLHGVLHYIISSVIQKFVLLILKRKTYNSHLAESTSPVQSMLDAYFPELIANFAASLCSDVILYPLETVLHRLHIQGTRTIIDNTDLGYEVLPINTQYEGMRDCINTIRQEEGVFGFYKGFGAVIIQYTLHAAVLQITKIIYSTLLQNNI
RUO
SLC25A46
Yes
Videos
SDS
Documents
Certificates