Learn More
Abnova™ Human SLC27A5 Partial ORF (NP_036386, 90 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010998-Q01.25ug
Additional Details : Weight : 0.00010kg
Descrizione
The protein encoded by this gene is an isozyme of very long-chain acyl-CoA synthetase (VLCS). It is capable of activating very long-chain fatty-acids containing 24- and 26-carbons. It is expressed in liver and associated with endoplasmic reticulum but not with peroxisomes. Its primary role is in fatty acid elongation or complex lipid synthesis rather than in degradation. This gene has a mouse ortholog. [provided by RefSeq]
Sequence: DVIFLAKILHLGLKIRGCLSRQPPDTFVDAFERRARAQPGRALLVWTGPGAGSVTFGELDARACQAAWALKAELGDPASLCAGEPTSpecifica
NP_036386 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVIFLAKILHLGLKIRGCLSRQPPDTFVDAFERRARAQPGRALLVWTGPGAGSVTFGELDARACQAAWALKAELGDPASLCAGEPT | |
RUO | |
SLC27A5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10998 | |
SLC27A5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACSB/ACSVL6/FACVL3/FATP5/FLJ22987/VLACSR/VLCS-H2/VLCSH2 | |
SLC27A5 | |
Recombinant | |
wheat germ expression system |