Learn More
Abnova™ Human SLC29A4 Partial ORF (NP_694979, 283 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00222962-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the SLC29 family and encodes a plasma membrane protein with 11 transmembrane helices. This protein catalyzes the reuptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It has been shown to transport several compounds, including serotonin, dopamine, and the neurotoxin 1-methyl-4-phenylpyridinium. Alternate transcriptional splice variants which encode the same protein have been characterized. [provided by RefSeq]
Sequence: VHHDVVAGDVHFEHPAPAPAPNESPKDSPAHEVTGSGGAYMRFDVPRPRVQRSWPTFRALLLHSpecifications
NP_694979 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.67kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VHHDVVAGDVHFEHPAPAPAPNESPKDSPAHEVTGSGGAYMRFDVPRPRVQRSWPTFRALLLH | |
RUO | |
SLC29A4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
222962 | |
SLC29A4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENT4/FLJ34923/PMAT | |
SLC29A4 | |
Recombinant | |
wheat germ expression system |