missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC2A2 Partial ORF (NP_000331.1, 32 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006514-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16185355

  • 6845.00 SEK / 25µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. [provided by RefSeq]

Sequence: NAPQQVIISHYRHVLGVPLDDRKAINNYVINSTDELPTISYSMNPKPTPWAEEETVAAAQLITMLWS
Specifications

Specifications

NP_000331.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.11kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NAPQQVIISHYRHVLGVPLDDRKAINNYVINSTDELPTISYSMNPKPTPWAEEETVAAAQLITMLWS
RUO
SLC2A2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6514
SLC2A2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GLUT2
SLC2A2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers