missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC6A5 Partial ORF (NP_004202, 294 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004202 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9152 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180066
|
Abnova™
H00009152-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170066
|
Abnova™
H00009152-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: TLFYLFASFVSVLPWGSCNNPWNTPECKDKTKLLLDSCVISDHPKIQIKNSTFCMTAYPNVTMVNFTSQANKTFVSGSEEYFKYFVLKISAGIEYPGEIRSpecifications
NP_004202 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLYT2/NET1 | |
SLC6A5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9152 | |
SLC6A5 (Human) Recombinant Protein (Q01) | |
TLFYLFASFVSVLPWGSCNNPWNTPECKDKTKLLLDSCVISDHPKIQIKNSTFCMTAYPNVTMVNFTSQANKTFVSGSEEYFKYFVLKISAGIEYPGEIR | |
RUO | |
SLC6A5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |