missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLFN5 Full-length ORF (AAH21238.1, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00162394-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178933

  • 5030.41 SEK / 25µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSLRIDVDTNFPECVVDAGKVTLGTQQRQEMDPRLREKQNEIILRAVCALLNSGGGIIKAEIENKGYNYERHGVGLDVPPIFRSHLDKMQKENHFLIFVKSWNTEAGVPLATLCSNLYHRERTSTDVMDSQEALAFLKCRTQTPTNINVSNSLGPQAAQGSVQYEGNINVSAAALFDRKRLQYLEKLNLPESTHVEFVMFSTDVSHCVKDRLPKCVSAFANTEGGYVFFGVHDETCQVIGCEKEKIDLTSLRASIDGCIKKLPVHHFCTQRPEIKYVLNFLEVHDKGALRGYVCAIKVEKFCCAVFAKVPSSWQVKDNRVRQLPTREWTAWMMEADPG
Specifications

Specifications

AAH21238.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC150611/MGC150612/MGC19764
SLFN5
Yes
wheat germ expression system
Liquid
162394
SLFN5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLRIDVDTNFPECVVDAGKVTLGTQQRQEMDPRLREKQNEIILRAVCALLNSGGGIIKAEIENKGYNYERHGVGLDVPPIFRSHLDKMQKENHFLIFVKSWNTEAGVPLATLCSNLYHRERTSTDVMDSQEALAFLKCRTQTPTNINVSNSLGPQAAQGSVQYEGNINVSAAALFDRKRLQYLEKLNLPESTHVEFVMFSTDVSHCVKDRLPKCVSAFANTEGGYVFFGVHDETCQVIGCEKEKIDLTSLRASIDGCIKKLPVHHFCTQRPEIKYVLNFLEVHDKGALRGYVCAIKVEKFCCAVFAKVPSSWQVKDNRVRQLPTREWTAWMMEADPG
RUO
SLFN5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates