Learn More
Abnova™ Human SMAP1 Full-length ORF (NP_068759.2, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00060682-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar to the mouse stromal membrane-associated protein-1. This similarity suggests that this human gene product is also a type II membrane glycoprotein involved in the erythropoietic stimulatory activity of stromal cells. Alternate splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MATRSCREKAQKLNEQHQLILSKLLREEDNKYCADCEAKGPRWASWNIGVFICIRCAGIHRNLGVHISRVKSVNLDQWTAEQIQCMQDMGNTKARLLYEANLPENFRRPQTDQAVEFFIRDKYEKKKYYDKNAIAITNKEKEKKKEEKKREKEPEKPAKPLTAEKLQKKDQQLEPKKSTSPKKAAEPTVDLLGLDGPAVAPVTNGNTTVPPLNDDLDIFGPMISNPLPATVMPPAQGTPSAPAAATLSTVTSGDLDLFTEQTTKSEEVAKKQLSKDSILSLYGTGTIQQQSTPGVFMGPTNIPFTSQAPAAFQGFPSMGVPVPAAPGLIGNVMGQSPSMMVGMPMPNGFMGNAQTGVMPLPQNVVGPQGGMVGQMGAPQSKFGLPQAQQPQWSLSQMNQQMAGMSISSATPTAGFGQPSSTTAGWSGSSSGQTLSTQLWKSpecifications
NP_068759.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ13159/FLJ42245/SMAP-1 | |
SMAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
60682 | |
SMAP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATRSCREKAQKLNEQHQLILSKLLREEDNKYCADCEAKGPRWASWNIGVFICIRCAGIHRNLGVHISRVKSVNLDQWTAEQIQCMQDMGNTKARLLYEANLPENFRRPQTDQAVEFFIRDKYEKKKYYDKNAIAITNKEKEKKKEEKKREKEPEKPAKPLTAEKLQKKDQQLEPKKSTSPKKAAEPTVDLLGLDGPAVAPVTNGNTTVPPLNDDLDIFGPMISNPLPATVMPPAQGTPSAPAAATLSTVTSGDLDLFTEQTTKSEEVAKKQLSKDSILSLYGTGTIQQQSTPGVFMGPTNIPFTSQAPAAFQGFPSMGVPVPAAPGLIGNVMGQSPSMMVGMPMPNGFMGNAQTGVMPLPQNVVGPQGGMVGQMGAPQSKFGLPQAQQPQWSLSQMNQQMAGMSISSATPTAGFGQPSSTTAGWSGSSSGQTLSTQLWK | |
RUO | |
SMAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |