missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SMYD5 Full-length ORF (NP_006053.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010322-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114892

  • 4055.00 SEK / 10µg
Estimated Shipment: 18-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MYCSAECRLAATEQYHQVLCPGPSQDDPLHPLNKLQEAWRSIHYPPETASIMLMARMVATVKQAKDKDRWIRLFSQFCNKTANEEEEIVHKLLGDKFKGQLELLRRLFTEALYEEAVSQWFTPDGFRSLFALVGTNGQGIGTSSLSQWVHACDTLELKPQDREQLDAFIDQLYKDIEAATGEFLNCEGSGLFVLQSCCNHSCVPNAETSFPENNFLLHVTALEDIKPGEEICISYLDCCQRERSRHSRHKILRENYLFVCSCPKCLAEADEPNVTSEEEEEEEEEEEGEPEDAELGDEMTDV
Specifications

Specifications

NP_006053.1
Liquid
10322
SMYD5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYCSAECRLAATEQYHQVLCPGPSQDDPLHPLNKLQEAWRSIHYPPETASIMLMARMVATVKQAKDKDRWIRLFSQFCNKTANEEEEIVHKLLGDKFKGQLELLRRLFTEALYEEAVSQWFTPDGFRSLFALVGTNGQGIGTSSLSQWVHACDTLELKPQDREQLDAFIDQLYKDIEAATGEFLNCEGSGLFVLQSCCNHSCVPNAETSFPENNFLLHVTALEDIKPGEEICISYLDCCQRERSRHSRHKILRENYLFVCSCPKCLAEADEPNVTSEEEEEEEEEEEGEPEDAELGDEMTDV
RUO
SMYD5
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NN8-4AG/RAI15/RRG1/ZMYND23
SMYD5
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers