Learn More
Abnova™ Human SNCAIP Full-length ORF (AAH94759.1, 1 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009627-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing several protein-protein interaction domains, including ankyrin-like repeats, a coiled-coil domain, and an ATP/GTP-binding motif. The encoded protein interacts with alpha-synuclein in neuronal tissue and may play a role in the formation of cytoplasmic inclusions and neurodegeneration. A mutation in this gene has been associated with Parkinson's disease. Alternatively spliced transcript variants encoding different isoforms of this gene have been described, but their full-length nature has yet to be determined. [provided by RefSeq]
Sequence: MTYLIQSHHSRRSQNCAEDVIRKTKTDQNGQLECVRWMVSETEAIAELSCSKDFPSLIHYAGCYGQEKILLWLLQFMQEQGISLDEVDQDGNSAVHVASQHGYLGCIQTLVEYGANVTMQNHAGEKPSQSAERQGHTLCSRYLVVVETCMSLASQVVKLTKQLKEQTVERVTLQNQLQQFLEAQKSEGKSLPSSPSSPSSPASRKSQWKSPDADDDSVAKSKPGVQEGIQVLGSLSASSRARPKAKDEDSDKILRQLLGKEISENVCTQEKLSLEFQDAQASSRNSKKIPLEKRELKLARLRQLMQRSLSESDTDSNNSEDPKTTPVRKADRPRPQPIVESVESMGSAESLHLMIKKHTLASGGRRFPFSIKASKSLDGHSPSPTSESSEPDLESQYPGSGSIPPNQPSGDPQQPSPDSTAAQKVATSPKSALKSPSSKRRTSQNLKLRVTFEEPVVQMEQPSLELNGEKDKDKGRTLQRTSTSNESGDQLKRPFGAFRSIMETLSGNQNNNNNYQAANQLKTSTLPLTSLGRKTDAKGNPASSASKGKNKAEMYSSCINLSSNMLIEEHLCNDTRHNDINRKMKKSYSIKHIAEPESKELFLSpecifications
AAH94759.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC39814/SYPH1 | |
SNCAIP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9627 | |
SNCAIP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTYLIQSHHSRRSQNCAEDVIRKTKTDQNGQLECVRWMVSETEAIAELSCSKDFPSLIHYAGCYGQEKILLWLLQFMQEQGISLDEVDQDGNSAVHVASQHGYLGCIQTLVEYGANVTMQNHAGEKPSQSAERQGHTLCSRYLVVVETCMSLASQVVKLTKQLKEQTVERVTLQNQLQQFLEAQKSEGKSLPSSPSSPSSPASRKSQWKSPDADDDSVAKSKPGVQEGIQVLGSLSASSRARPKAKDEDSDKILRQLLGKEISENVCTQEKLSLEFQDAQASSRNSKKIPLEKRELKLARLRQLMQRSLSESDTDSNNSEDPKTTPVRKADRPRPQPIVESVESMGSAESLHLMIKKHTLASGGRRFPFSIKASKSLDGHSPSPTSESSEPDLESQYPGSGSIPPNQPSGDPQQPSPDSTAAQKVATSPKSALKSPSSKRRTSQNLKLRVTFEEPVVQMEQPSLELNGEKDKDKGRTLQRTSTSNESGDQLKRPFGAFRSIMETLSGNQNNNNNYQAANQLKTSTLPLTSLGRKTDAKGNPASSASKGKNKAEMYSSCINLSSNMLIEEHLCNDTRHNDINRKMKKSYSIKHIAEPESKELFL | |
RUO | |
SNCAIP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |