missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SNRK Full-length ORF (AAH71567.1, 1 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH71567.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54861 |
Molecular Weight (g/mol) | 110.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170833
|
Abnova™
H00054861-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180833
|
Abnova™
H00054861-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SNRK is a member of the sucrose nonfermenting (SNF)-related kinase family of serine/threonine kinases (Kertesz et al., 2002 [PubMed 12234663]).[supplied by OMIM]
Sequence: MAGFKRGYDGKIAGLYDLDKTLGRGHFAVVKLARHVFTGEKVAVKVIDKTKLDTLATGHLFQEVRCMKLVQHPNIVRLYEVIDTQTKLYLILELGDGGDMFDYIMKHEEGLNEDLAKKYFAQIVHAISYCHKLHVVHRDLKPENVVFFEKQGLVKLTDFGFSNKFQPGKKLTTSCGSLAYSAPEILLGDEYDAPAVDIWSLGVILFMLVCGQPPFQEANDSETLTMIMDCKYTVPSHVSKECKDLITRMLQRDPKRRAYLEEIENHPWLQGVDPSPATKYNIPLVSYKNLSEEEHNSIIQRMVLGDIADRDAIVEALETNRYNHITATYFLLAERILREKQEKEIQTRSASPSNIKAQFRQSWPTKIDVPQDLEDDLTATPLSHATVPQSPARAADSVLNGHRSKGLCDSAKKDDLPELAGPALSTVPPASLKPTASGRKCLFRVEEDEEEDEEDKKPMSLSTQVVLRRKPSVTNRLTSRKSAPVVNQIFEEGESDDEFDMDENLPPKLSRLKMNIASPGTVHKRYHRRKSQGRGSSCSSSETSDDDSESRRRLDKDSGFTYSWHRRDSSEGPPGSEGDGGGQSKPSNASGGVDKASPSENNAGGGSPSSGSGGNPTNTSGTTRRCAGPSNSMQLASRSAGELVESLKLMSLCLGSQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIRSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVISpecifications
AAH71567.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAGFKRGYDGKIAGLYDLDKTLGRGHFAVVKLARHVFTGEKVAVKVIDKTKLDTLATGHLFQEVRCMKLVQHPNIVRLYEVIDTQTKLYLILELGDGGDMFDYIMKHEEGLNEDLAKKYFAQIVHAISYCHKLHVVHRDLKPENVVFFEKQGLVKLTDFGFSNKFQPGKKLTTSCGSLAYSAPEILLGDEYDAPAVDIWSLGVILFMLVCGQPPFQEANDSETLTMIMDCKYTVPSHVSKECKDLITRMLQRDPKRRAYLEEIENHPWLQGVDPSPATKYNIPLVSYKNLSEEEHNSIIQRMVLGDIADRDAIVEALETNRYNHITATYFLLAERILREKQEKEIQTRSASPSNIKAQFRQSWPTKIDVPQDLEDDLTATPLSHATVPQSPARAADSVLNGHRSKGLCDSAKKDDLPELAGPALSTVPPASLKPTASGRKCLFRVEEDEEEDEEDKKPMSLSTQVVLRRKPSVTNRLTSRKSAPVVNQIFEEGESDDEFDMDENLPPKLSRLKMNIASPGTVHKRYHRRKSQGRGSSCSSSETSDDDSESRRRLDKDSGFTYSWHRRDSSEGPPGSEGDGGGQSKPSNASGGVDKASPSENNAGGGSPSSGSGGNPTNTSGTTRRCAGPSNSMQLASRSAGELVESLKLMSLCLGSQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIRSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVI | |
RUO | |
SNRK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54861 | |
SNRK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779A1866/FLJ20224/HSNFRK/KIAA0096 | |
SNRK | |
Recombinant | |
wheat germ expression system |