missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SNRK Partial ORF (NP_060189, 656 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054861-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
SNRK is a member of the sucrose nonfermenting (SNF)-related kinase family of serine/threonine kinases (Kertesz et al., 2002 [PubMed 12234663]).[supplied by OMIM]
Sequence: SQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIKSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVISpecifications
NP_060189 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIKSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVI | |
RUO | |
SNRK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54861 | |
SNRK (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779A1866/FLJ20224/HSNFRK/KIAA0096 | |
SNRK | |
Recombinant | |
wheat germ expression system |