Learn More
Abnova™ Human SORT1 Partial ORF (NP_002950, 203 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006272-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is a multi-ligand type-1 receptor with similarity to the yeast carboxypeptidase Y sorting receptor Vps10 protein. The encoded protein, a trans-Golgi network (TGN) transmembrane protein, binds a number of unrelated ligands that participate in a wide range of cellular processes; however, it lacks the typical features of a signalling receptor. In the TGN, furin mediates the activation of the mature binding form. The encoded protein consists of a large luminal domain, a single transmembrane segment and short C-terminal cytoplasmic tail. The luminal domain contains a cysteine-rich region similar to two corresponding segments in the yeast Vps10p; the cytoplasmic tail is similar to the corresponding segment of the cation-independent mannose 6-phosphate receptor and the tail also interacts with the VHS domains of GGA (Golgi-associated, gamma-adaptin homologous, ARF-interacting) proteins. [provided by RefSeq]
Sequence: FAKNFVQTDLPFHPLTQMMYSPQNSDYLLALSTENGLWVSKNFGGKWEEIHKAVCLAKWGSDNTIFFTTYANGSCKADLGALELWRTSDLGKSFKTISpecifications
NP_002950 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FAKNFVQTDLPFHPLTQMMYSPQNSDYLLALSTENGLWVSKNFGGKWEEIHKAVCLAKWGSDNTIFFTTYANGSCKADLGALELWRTSDLGKSFKTI | |
RUO | |
SORT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6272 | |
SORT1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Gp95/NT3 | |
SORT1 | |
Recombinant | |
wheat germ expression system |