Learn More
Abnova™ Human SOX6 Partial ORF (NP_201583, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_201583 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55553 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188196
|
Abnova™
H00055553-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178196
|
Abnova™
H00055553-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the D subfamily of sex determining region y-related transcription factors that are characterized by a conserved DNA-binding domain termed the high mobility group box and by their ability to bind the minor groove of DNA. The encoded protein is a transcriptional activator that is required for normal development of the central nervous system, chondrogenesis and maintenance of cardiac and skeletal muscle cells. The encoded protein interacts with other family members to cooperatively activate gene expression. Alternative splicing results in multiple transcript variants
Sequence: MSSKQATSPFACAADGEDAMTQDLTSREKEEGSDQHVASHLPLHPIMHNKPHSEELPTLVSTIQQDADWDSVLSSQQRMESENNKLCSLYSFRNTSTSPHKPDEGSRDRSpecifications
NP_201583 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSSOX6 | |
SOX6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55553 | |
SOX6 (Human) Recombinant Protein (Q01) | |
MSSKQATSPFACAADGEDAMTQDLTSREKEEGSDQHVASHLPLHPIMHNKPHSEELPTLVSTIQQDADWDSVLSSQQRMESENNKLCSLYSFRNTSTSPHKPDEGSRDR | |
RUO | |
SOX6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |