Learn More
Abnova™ Human SP7 Partial ORF (NP_690599, 87 a.a. - 162 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00121340-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation (Gao et al., 2004 [PubMed 15474293]).[supplied by OMIM]
Sequence: GYANDYPPFSHSFPGPTGTQDPGLLVPKGHSSSDCLPSVYTSLDMTHPYGSWYKAGIHAGISPGPGNTPTPWWDMHSpecifications
NP_690599 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GYANDYPPFSHSFPGPTGTQDPGLLVPKGHSSSDCLPSVYTSLDMTHPYGSWYKAGIHAGISPGPGNTPTPWWDMH | |
RUO | |
SP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
121340 | |
SP7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126598/OSX/osterix | |
SP7 | |
Recombinant | |
wheat germ expression system |