Learn More
Abnova™ Human SPACA1 Full-length ORF (NP_112222.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_112222.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81833 |
Molecular Weight (g/mol) | 58.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16175253
|
Abnova™
H00081833-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16185253
|
Abnova™
H00081833-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein expressed by this gene is recognized by anti-sperm antibodies from infertile males. Furthermore, antibodies generated against the recombinant protein block in vitro fertilization. This protein localizes to the acrosomal membrane of spermatids and mature spermatozoa where it is thought to play a role in acrosomal morphogenesis and in sperm-egg binding and fusion, respectively. [provided by RefSeq]
Sequence: MSPRGTGCSAGLLMTVGWLLLAGLQSARGTNVTAAVQDAGLAHEGEGEEETENNDSETAENYAPPETEDVSNRNVVKEVEFGMCTVTCGIGVREVILTNGCPGGESKCVVRVEECRGPTDCGWGKPISESLESVRLACIHTSPLNRFKYMWKLLRQDQQSIILVNDSAILEVRKESHPLAFECDTLDNNEIVATIKFTVYTSSELQMRRSSLPATDAALIFVLTIGVIICVFIIFLLIFIIINWAAVKAFWGAKASTPEVQSEQSSVRYKDSTSLDQLPTEMPGEDDALSEWNESpecifications
NP_112222.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSPRGTGCSAGLLMTVGWLLLAGLQSARGTNVTAAVQDAGLAHEGEGEEETENNDSETAENYAPPETEDVSNRNVVKEVEFGMCTVTCGIGVREVILTNGCPGGESKCVVRVEECRGPTDCGWGKPISESLESVRLACIHTSPLNRFKYMWKLLRQDQQSIILVNDSAILEVRKESHPLAFECDTLDNNEIVATIKFTVYTSSELQMRRSSLPATDAALIFVLTIGVIICVFIIFLLIFIIINWAAVKAFWGAKASTPEVQSEQSSVRYKDSTSLDQLPTEMPGEDDALSEWNE | |
RUO | |
SPACA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
81833 | |
SPACA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC32952/SAMP32 | |
SPACA1 | |
Recombinant | |
wheat germ expression system |