Learn More
Abnova™ Human SPIB Full-length ORF (NP_003112.2, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006689-P01.25ug
Additional Details : Weight : 0.00010kg
Description
SPI1 (MIM 165170) and SPIB are members of a subfamily of ETS (see ETS1; MIM 164720) transcription factors. ETS proteins share a conserved ETS domain that mediates specific DNA binding. SPIB and SPI1 bind to a purine-rich sequence, the PU box (5-prime-GAGGAA-3-prime).[supplied by OMIM]
Sequence: MLALEAAQLDGPHFSCLYPDGVFYDLDSCKHSSYPDSEGAPDSLWDWTVAPPVPATPYEAFDPAAAAFSHPQAAQLCYEPPTYSPAGNLELAPSLEAPGPGLPAYPTENFASQTLVPPAYAPYPSPVLSEEEDLPLDSPALEVSDSESDEALVAGPEGKGSEAGTRKKLRLYQFLLGLLTRGDMRECVWWVEPGAGVFQFSSKHKELLARRWGQQKGNRKRMTYQKLARALRNYAKTGEIRKVKRKLTYQFDSALLPAVRRASpecifications
NP_003112.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPI-B | |
SPIB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6689 | |
SPIB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLALEAAQLDGPHFSCLYPDGVFYDLDSCKHSSYPDSEGAPDSLWDWTVAPPVPATPYEAFDPAAAAFSHPQAAQLCYEPPTYSPAGNLELAPSLEAPGPGLPAYPTENFASQTLVPPAYAPYPSPVLSEEEDLPLDSPALEVSDSESDEALVAGPEGKGSEAGTRKKLRLYQFLLGLLTRGDMRECVWWVEPGAGVFQFSSKHKELLARRWGQQKGNRKRMTYQKLARALRNYAKTGEIRKVKRKLTYQFDSALLPAVRRA | |
RUO | |
SPIB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |