Learn More
Invitrogen™ Human SRC1 (aa 8-24) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP044
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human SRC-1 (residues 8-24) Synthetic Peptide, PEP-044, from Invitrogen.
PEP-044 is a 17 amino acid synthetic peptide whose sequence corresponds to the N-terminal residues 8-24 of human SRC-1. The sequence of this peptide is (amino to carboxy terminus): S8-S-D-P-A-N-P-D-S-H-K-R-K-G-S-P-C24This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SRC-1, catalog # PA1-840. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
8648 | |
purified | |
Human SRC1 (aa 8-24) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q15788 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NCOA1 | |
No preservative | |
50 μg | |
S8 - S - D - P - A - N - P - D - S - H - K - R - K - G - S - P - C24 |