missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SSX2 Full-length ORF (AAH02818, 1 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006757-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121042

  • 4055.00 SEK / 10µg
Estimated Shipment: 07-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This translocation results in the fusion of the synovial sarcoma translocation gene on chromosome 18 to one of the SSX genes on chromosome X. The encoded hybrid proteins are probably responsible for transforming activity. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MNGDDAFARRPTVGAQIPEKIQKAFDDIAKYFSKEEWEKMKASEKIFYVYMKRKYEAMTKLGFKATLPPFMCNKRAEDFQGNDLDNDPNRGNQVERPQMTFGRLQGISPKIMPKKPAEEGNDSEEVPEASGPQNDGKELCPPGKPTTSEKIHERSGNREAQEKEERRGTAHRWSSQNTHNIGRFSLSTSMGAVHGTPKTITHNRDPKGGNMPGPTDCVRENSW
Specifications

Specifications

AAH02818
Liquid
6757
SSX2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGDDAFARRPTVGAQIPEKIQKAFDDIAKYFSKEEWEKMKASEKIFYVYMKRKYEAMTKLGFKATLPPFMCNKRAEDFQGNDLDNDPNRGNQVERPQMTFGRLQGISPKIMPKKPAEEGNDSEEVPEASGPQNDGKELCPPGKPTTSEKIHERSGNREAQEKEERRGTAHRWSSQNTHNIGRFSLSTSMGAVHGTPKTITHNRDPKGGNMPGPTDCVRENSW
RUO
SSX2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.27kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HD21/HOM-MEL-40/MGC119055/MGC15364/MGC3884/SSX
SSX2
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers