missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST6GALNAC2 Full-length ORF (AAH40455.1, 1 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010610-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135412

  • 3343.96 SEK / 10µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).[supplied by OMIM]

Sequence: MGLPRGSFFWLLLLLTAACSGLLFALYFSAVQRYPGPAAGARDTTSFEAFFQSKASNSWTGKGQACRHLLHLAIQRHPHFRGLFNLSIPVLLWGDLFTPALWDRLSQHKAPYGWRGLSHQVIASTLSLLNGSESAKLFAPPRDTPPKCIRCAVVGNGGILNGSRQGPNIDAHDYVFRLNGAVIKGFERDVGTKTSFYGFTVNTMKNSLVSYWNLGFTSVPQGQDLQYIFIPSDIRDYVMLRSAILGVPVPEGLDKGDRPHAYFGPEASASKFKLLHPDFISYLTERFLKSKLINTHFGDLYMPSTGALMLLTALHTCDQVSAYGFITSNYWKFSDHYFERKMKPLIFYANHDLSLEAALWRDLHKAGILQLYQR
Specifications

Specifications

AAH40455.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ45660/SAITL1/SIAT7/SIAT7B/SIATL1/ST6GalNAII/STHM
ST6GALNAC2
Yes
wheat germ expression system
Liquid
10610
ST6GALNAC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLPRGSFFWLLLLLTAACSGLLFALYFSAVQRYPGPAAGARDTTSFEAFFQSKASNSWTGKGQACRHLLHLAIQRHPHFRGLFNLSIPVLLWGDLFTPALWDRLSQHKAPYGWRGLSHQVIASTLSLLNGSESAKLFAPPRDTPPKCIRCAVVGNGGILNGSRQGPNIDAHDYVFRLNGAVIKGFERDVGTKTSFYGFTVNTMKNSLVSYWNLGFTSVPQGQDLQYIFIPSDIRDYVMLRSAILGVPVPEGLDKGDRPHAYFGPEASASKFKLLHPDFISYLTERFLKSKLINTHFGDLYMPSTGALMLLTALHTCDQVSAYGFITSNYWKFSDHYFERKMKPLIFYANHDLSLEAALWRDLHKAGILQLYQR
RUO
ST6GALNAC2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates