Learn More
Abnova™ Human STAB2 Partial ORF (NP_060034, 367 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_060034 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55576 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16108216
|
Abnova™
H00055576-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16118216
|
Abnova™
H00055576-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a large, transmembrane receptor protein which may function in angiogenesis, lymphocyte homing, cell adhesion, or receptor scavenging. The protein contains 7 fasciclin, 15 epidermal growth factor (EGF)-like, and 2 laminin-type EGF-like domains as well as a C-type lectin-like hyaluronan-binding Link module. The protein is primarily expressed on sinusoidal endothelial cells of liver, spleen, and lymph node. The receptor has been shown to bind and endocytose ligands such as hyaluronan, low density lipoprotein, Gram-positive and Gram-negative bacteria, and advanced glycosylation end products. Supporting its possible role as a scavenger receptor, the protein has been shown to cycle between the plasma membrane and lysosomes. [provided by RefSeq]
Sequence: ERLRELNTEPRGKWQGRLTSFISLLDKAYAWPLSKLGPFTVLLPTDKGLKGFNVNELLVDNKAAQYFVKLHIIAGQMNIEYMNNTDMFYTLTGKSGEIFSpecifications
NP_060034 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434E0321/FEEL-2/FELE-2/FELL/FELL-2/FEX2/HARE/STAB-2 | |
STAB2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55576 | |
STAB2 (Human) Recombinant Protein (Q01) | |
ERLRELNTEPRGKWQGRLTSFISLLDKAYAWPLSKLGPFTVLLPTDKGLKGFNVNELLVDNKAAQYFVKLHIIAGQMNIEYMNNTDMFYTLTGKSGEIF | |
RUO | |
STAB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |