Learn More
Human STAM (STAM1) knockout HEK-293T cell lysate, 1 Kit
Product Code. 32155578 Shop All Abcam Products
Product Code. 32155578 Supplier Abcam Supplier No. ab2591581Kit
 Restricted Item
Beräknad leverans: 30-11-2026
Human STAM (STAM1) knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: STAM (STAM1) knockout HEK-293T