missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Human STAT1 (NP_009330) VersaClone cDNA

Produktkod. 18711329
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18711329 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18711329 Leverantör R&D Systems™ Leverantörsnummer RDC1455

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human STAT1 (NP_009330) VersaClone cDNA clone contains the complete ORF for STAT1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The STAT1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Beställningsinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifikationer

Gen STAT1
Tillträdesnummer NP_009330
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.128
Infoga typ 2265bp
Gen-ID (Entrez) 6772
Gene Alias CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Human
Kvantitet 10 μg

For Reasearch Use Only. Not for use in Humans.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.