missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STCH Full-length ORF (NP_008879.3, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_008879.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6782 |
Molecular Weight (g/mol) | 78.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123268
|
Abnova™
H00006782-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133268
|
Abnova™
H00006782-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein. [provided by RefSeq]
Sequence: MAREMTILGSAVLTLLLAGYLAQQYLPLPTPKVIGIDLGTTYCSVGVFFPGTGKVKVIPDENGHISIPSMVSFTDNDVYVGYESVELADSNPQNTIYDAKRFIGKIFTAEELEAEIGRYPFKVLNKNGMVEFSVTSNETITVSPEYVGSRLLLKLKEMAEAYLGMPVANAVISVPAEFDLKQRNSTIEAANLAGLKILRVINEPTAAAMAYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDHRVNSGFGRGLSDKKSGESQVLFETEISRKLFDTLNEDLFQKILVPIQQVLKEGHLEKTEIDEVVLVGGSTRIPRIRQVIQEFFGKDPNTSVDPDLAVVTGVAIQAGIDGGSWPLQVSALEIPNKHLQKTNFNSpecifications
NP_008879.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAREMTILGSAVLTLLLAGYLAQQYLPLPTPKVIGIDLGTTYCSVGVFFPGTGKVKVIPDENGHISIPSMVSFTDNDVYVGYESVELADSNPQNTIYDAKRFIGKIFTAEELEAEIGRYPFKVLNKNGMVEFSVTSNETITVSPEYVGSRLLLKLKEMAEAYLGMPVANAVISVPAEFDLKQRNSTIEAANLAGLKILRVINEPTAAAMAYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDHRVNSGFGRGLSDKKSGESQVLFETEISRKLFDTLNEDLFQKILVPIQQVLKEGHLEKTEIDEVVLVGGSTRIPRIRQVIQEFFGKDPNTSVDPDLAVVTGVAIQAGIDGGSWPLQVSALEIPNKHLQKTNFN | |
RUO | |
HSPA13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6782 | |
STCH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STCH | |
HSPA13 | |
Recombinant | |
wheat germ expression system |