Läs mer
Invitrogen™ Human Stem Cell Factor (SCF) Synthetic Peptide
Kvantitet:
50 μg
Förpackningsstorlek:
50µg
Beskrivning
SCF Synthetic Peptide, PEP-0860, from Invitrogen.
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
Specifikationer
Specifikationer
| Gen-ID (Entrez) | 4254 |
| Art | Human |
| Reningsmetod | purified |
| Tillträdesnummer | P21583 |
| Vanligt namn | Human Stem Cell Factor (SCF) |
| Koncentration | 200 μg/mL |
| Innehåll och lagring | -20°C |
| Beskrivning | Synthetic Peptide |
| Formatera | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Visa mer |