missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STK38 Partial ORF (AAH12085, 365 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011329-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184086

  • 3343.96 SEK / 10µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: EHRIGAPGVEEIKSNSFFEGVDWEHIRERPAAISIEIKSIDDTSNFDEFPESDILKPTVATSNHPETDYKNKDWVFINYTYKRFEGLTARGAIPSYMKAAK
Specifications

Specifications

AAH12085
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EHRIGAPGVEEIKSNSFFEGVDWEHIRERPAAISIEIKSIDDTSNFDEFPESDILKPTVATSNHPETDYKNKDWVFINYTYKRFEGLTARGAIPSYMKAAK
RUO
STK38
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
11329
STK38 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NDR/NDR1
STK38
Yes
Videos
SDS
Documents
Certificates