missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STXBP3 Full-length ORF (AAH38099.1, 1 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH38099.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6814 |
Molecular Weight (g/mol) | 94.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121122
|
Abnova™
H00006814-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131122
|
Abnova™
H00006814-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MAPPVAERGLKSVVWQKIKATVFDDCKKEGEWKIMLLDEFTTKLLASCCKMTDLLEEGITVVENIYKNREPVRQMKALYFITPTSKSVDCFLHDFASKSENKYRAAYIYFTDFCPDNLFNKIKASCSKSIRRCKEINISFIPHESQVYTRDVPDAFYYCYSPDPGNAKGKDAIMETMADQIVTVCATLDENPGVRYKSKPLDNASKLAQLVEKKLEDYYKIDEKSLIKGKTHSQLLIIDRGFDPVSTVLHELTFQAMAYDLLPIENDTYKYKTDGKEKEAILEEEDDLWVRIRHRHIAVVLEEIPKLMKEISSTKKATEGKTSLSALTQLMKKMPHFRKQITKQVVHLNLAEDCMNKFKLNIEKLCKTEQDLALGTDAEGQKVKDSMRVLLPVLLNKNHDNCDKIRAILLYIFSINGTTEENLDRLIQNVKIENESDMIRNWSYLGVPIVPQSQQGKPLRKDRSAEETFQLSRWTPFIKDIMEDAIDNRLDSKEWPYCSQCPAVWNGSGAVSARQKPRANYLEDRKNGSKLIVFVIGGITYSEVRCAYEVSQAHKSCEVIIGSTHVLTPKKLLDDIKMLNKPKDKVSLIKDESpecifications
AAH38099.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPPVAERGLKSVVWQKIKATVFDDCKKEGEWKIMLLDEFTTKLLASCCKMTDLLEEGITVVENIYKNREPVRQMKALYFITPTSKSVDCFLHDFASKSENKYRAAYIYFTDFCPDNLFNKIKASCSKSIRRCKEINISFIPHESQVYTRDVPDAFYYCYSPDPGNAKGKDAIMETMADQIVTVCATLDENPGVRYKSKPLDNASKLAQLVEKKLEDYYKIDEKSLIKGKTHSQLLIIDRGFDPVSTVLHELTFQAMAYDLLPIENDTYKYKTDGKEKEAILEEEDDLWVRIRHRHIAVVLEEIPKLMKEISSTKKATEGKTSLSALTQLMKKMPHFRKQITKQVVHLNLAEDCMNKFKLNIEKLCKTEQDLALGTDAEGQKVKDSMRVLLPVLLNKNHDNCDKIRAILLYIFSINGTTEENLDRLIQNVKIENESDMIRNWSYLGVPIVPQSQQGKPLRKDRSAEETFQLSRWTPFIKDIMEDAIDNRLDSKEWPYCSQCPAVWNGSGAVSARQKPRANYLEDRKNGSKLIVFVIGGITYSEVRCAYEVSQAHKSCEVIIGSTHVLTPKKLLDDIKMLNKPKDKVSLIKDE | |
RUO | |
STXBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6814 | |
STXBP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MUNC18-3/MUNC18C/PSP/UNC-18C | |
STXBP3 | |
Recombinant | |
wheat germ expression system |